Methyl-2-(bromomethyl)-3-nitrobenzoate ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Methyl 2-(bromomethyl)-3-nitrobenzoate
M2422-1G
M2422-5G
TCIAM2422-1GEA
455
SEK
TCIAM2422-1G
TCIAM2422-5G
Methyl-2-(bromomethyl)-3-nitrobenzoate ≥98.0%
Methyl-2-(bromomethyl)-3-nitrobenzoate
Formel:
C₉H₈BrNO₄ MW: 274,07 g/mol |
MDL Number:
MFCD04114315 CAS nummer: 98475-07-1 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...