erythro-3-Methoxy-2-methyl-4-phenylbutyric acid ≥98.0%

Leverantör: TCI
Varning

M2415-100MG M2415-500MG
TCIAM2415-100MGEA 1325 SEK
TCIAM2415-100MG TCIAM2415-500MG
erythro-3-Methoxy-2-methyl-4-phenylbutyric acid ≥98.0%
erythro-3-Methoxy-2-methyl-4-phenylbutyric acid
Formel: C₁₂H₁₆O₃
Storage Temperature: Frys
CAS nummer: 833483-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb1c8ea35376e5b68e5d857e0c392b90d5d59d32&vid=fb1c8ea35376e5b68e5d857e0c392b90d5d59d32&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR