2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde ≥97.0%

Leverantör: TCI
Varning

M2404-25G M2404-100G
TCIAM2404-25GEA 325 SEK
TCIAM2404-25G TCIAM2404-100G
2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde ≥97.0%
2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde
Formel: C₁₁H₁₂O₃
Kokpunkt: 154°C
MDL Number: MFCD00067053
CAS nummer: 1205-17-0
EINECS: 214-881-6
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5b4c585f80d7acb2c99d4fe9ff83e7ce230794d&vid=dd088c959241e5cb24fadad5868953525993ba0f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR