Mizoribine ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-hydroxy-1H-imidazole-4-carboxamide
M2399-50MG
M2399-250MG
TCIAM2399-50MGEA
805
SEK
TCIAM2399-50MG
TCIAM2399-250MG
Mizoribine ≥98.0%
Mizoribine
Formel:
C₉H₁₃N₃O₆ MW: 259,22 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00057221 CAS nummer: 50924-49-7 Merck Index: 14,06222 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 15.50 to 16.70 % |
Specific rotation [a]20/D | -25.0 to -27.0 deg (C=2,H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...