NSC 66811 ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
NSC66811, 2-Methyl-7-[phenyl(phenylamino)methyl]-8-quinolinol, 2-Methyl-7-(phenyl(phenylamino)methyl)quinolin-8-ol, 8-Hydroxy-2-methyl-7-[phenyl(phenylamino)methyl]quinoline
M2390-25MG
M2390-100MG
TCIAM2390-25MGEA
1675
SEK
TCIAM2390-25MG
TCIAM2390-100MG
NSC 66811 ≥98.0%
NSC 66811
Formel:
C₂₃H₂₀N₂O MW: 340,43 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD08823776 CAS nummer: 6964-62-1 |
Specifikation testresultat
Appearance | White to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 143.0 to 147.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...