Metamitron ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5-one
M2388-5G
M2388-25G
TCIAM2388-5GEA
485
SEK
TCIAM2388-5G
TCIAM2388-25G
Metamitron ≥98.0% (by GC, titration analysis)
Metamitron
Formel:
C₁₀H₁₀N₄O MW: 202,22 g/mol |
MDL Number:
MFCD00055524 CAS nummer: 41394-05-2 EINECS: 255-349-3 UN: 3077 ADR: 9,III Merck Index: 14,05924 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...