Methyl-2-bromohexanoate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl 2-bromohexanoate
M2378-25G
M2378-250G
TCIAM2378-25GEA
215
SEK
TCIAM2378-25G
TCIAM2378-250G
Methyl-2-bromohexanoate ≥98.0%
Methyl-2-bromohexanoate
Formel:
C₇H₁₃BrO₂ MW: 209,08 g/mol Kokpunkt: 94°C Flampunkt: 69°C |
MDL Number:
MFCD00045038 CAS nummer: 5445-19-2 EINECS: 226-643-9 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...