3-Nitro-4-picoline ≥97.0%

Leverantör: TCI
Varning

M2353-1G M2353-5G
TCIAM2353-1GEA 470 SEK
TCIAM2353-1G TCIAM2353-5G
3-Nitro-4-picoline ≥97.0%
3-Nitro-4-picoline
Formel: C₆H₆N₂O₂
Kokpunkt: 110°C
MDL Number: MFCD00051829
CAS nummer: 5832-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=895cfed8a0ef55a2bf5f588a5d316fd59fa1043d&vid=895cfed8a0ef55a2bf5f588a5d316fd59fa1043d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR