Methyl-4-(cyanomethyl)benzoate ≥95.0%
Leverantör: TCI
|
Varning
|
M2341-5G
M2341-25G
TCIAM2341-5GEA
990
SEK
TCIAM2341-5G
TCIAM2341-25G
Methyl-4-(cyanomethyl)benzoate ≥95.0%
Methyl-4-(cyanomethyl)benzoate
Formel:
C₁₀H₉NO₂ Kokpunkt: 112°C |
MDL Number:
MFCD00060695 CAS nummer: 76469-88-0 |
|
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...