EMCA (6-Maleimidohexanoic acid) ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
EMCA
M2338-1G
M2338-5G
TCIAM2338-1GEA
685
SEK
TCIAM2338-1G
TCIAM2338-5G
EMCA (6-Maleimidohexanoic acid) ≥98.0% (by GC, titration analysis)
EMCA (6-Maleimidohexanoic acid)
Formel:
C₁₀H₁₃NO₄ MW: 211,22 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00043140 CAS nummer: 55750-53-3 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 86.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...