4-(2-Methyl-4-thiazolyl)phenol ≥98.0% for biochemical research

Leverantör: TCI

Synonyms: 4-(2-Methyl-4-thiazolyl)phenol [for Biochemical Research]

M2325-100MG M2325-1G
TCIAM2325-100MGEA 335 SEK
TCIAM2325-100MG TCIAM2325-1G
4-(2-Methyl-4-thiazolyl)phenol ≥98.0% for biochemical research
4-(2-Methyl-4-thiazolyl)phenol
Formel: C₁₀H₉NOS
MDL Number: MFCD02642878
CAS nummer: 30686-73-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a06575890278672de463e455e623cb6cbfda3800&vid=faf6fbcdf020824c9e2c276768d56d90e5f1978e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (HPLC) min. 98.0 area%
Melting point 211.0 to 215.0 deg-C
Functionality test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR