Mitomycin C ≥98.0%
Leverantör: TCI
Fara
Synonyms: Mitomycin, Ametycin
M2320-10MG M2320-50MG
TCIAM2320-10MGEA 935 SEK
TCIAM2320-10MG TCIAM2320-50MG
Mitomycin C ≥98.0%
Mitomycin C
Formel: C₁₅H₁₈N₄O₅
MW: 334.33 g/mol
Smältpunkt: 64°C
Storage Temperature: Kylskåp
MDL Number: MFCD00078109
CAS nummer: 50-07-7
EINECS: 200-008-6
UN: 2811
ADR: 6.1 II
Merck Index: 14,06215
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9f718ca3f6067f36130655150f7562f770591e4&vid=d3b45ceb94edb9f7c5e6858897ad091c6198433b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 16.4 to 17.0 %