Methyl malonyl chloride ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: Methyl 3-chloro-3-oxopropanoate

M2315-5G M2315-25G
TCIAM2315-5GEA 525 SEK
TCIAM2315-5G TCIAM2315-25G
Methyl malonyl chloride ≥97.0% (by GC, titration analysis)
Methyl malonyl chloride
Formel: C₄H₅ClO₃
MW: 136,53 g/mol
Kokpunkt: 59°C
Flampunkt: 80°C
Storage Temperature: Kylskåp
MDL Number: MFCD00013657
CAS nummer: 37517-81-0
EINECS: 253-540-6
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a6f3cfd69ccc2c8f4dcf6fd818226b63bbd77fe&vid=2a6f3cfd69ccc2c8f4dcf6fd818226b63bbd77fe&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR