Methyl trans-6-octadecenoate ≥97.0%
Leverantör: TCI
M2310-100MG M2310-500MG
TCIAM2310-100MGEA 1250 SEK
TCIAM2310-100MG TCIAM2310-500MG
Methyl trans-6-octadecenoate ≥97.0%
Methyl trans-6-octadecenoate
Formel: C₁₉H₃₆O₂
Kokpunkt: 165°C
Smältpunkt: 19°C
Storage Temperature: Frys
MDL Number: MFCD00069607
CAS nummer: 14620-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e0c8ca6b7ac3cb5b21552586368e12777c875cc&vid=8e0c8ca6b7ac3cb5b21552586368e12777c875cc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %