O-Methyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI

Synonyms: (S)-2-Amino-3-(4-methoxyphenyl)propionic Acid, H-Tyr(Me)-OH, 4-Methoxy-L-phenylalanine

M2276-1G
TCIAM2276-1GEA 535 SEK
TCIAM2276-1G
O-Methyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)
O-Methyl-L-tyrosine
Formel: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00002604
CAS nummer: 6230-11-1
EINECS: 228-333-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e05bf60e02cb539ec55dd54736d336aaa62fbcda&vid=e05bf60e02cb539ec55dd54736d336aaa62fbcda&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -7.0 to -10.0 deg (C=2、1mol/LHCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR