2-Methacryloyloxy-2-methyladamantane ≥97.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: Methacrylic Acid 2-Methyl-2-adamantyl Ester, 2-Methyl-2-adamantyl Methacrylate, 2-Methyladamantan-2-yl methacrylate
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
M2260-5G M2260-1G
TCIAM2260-5GEA 2375 SEK
TCIAM2260-5G TCIAM2260-1G
2-Methacryloyloxy-2-methyladamantane ≥97.0% stabilised
2-Methacryloyloxy-2-methyladamantane
Formel: C₁₅H₂₂O₂
MW: 234,34 g/mol
Flampunkt: 120°C
Storage Temperature: Frys
MDL Number: MFCD04114354
CAS nummer: 177080-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab15e21b8289e0380d75ce355bc3e9fbb9ffe737&vid=ab15e21b8289e0380d75ce355bc3e9fbb9ffe737&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %