2-Methylpyridine-5-boronic acid hydrochloride (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Methyl-3-pyridylboronic acid hydrochloride

M2253-1G M2253-5G
TCIAM2253-1GEA 1150 SEK
TCIAM2253-1G TCIAM2253-5G
2-Methylpyridine-5-boronic acid hydrochloride (contains varying amounts of Anhydride)
2-Methylpyridine-5-boronic acid hydrochloride
Formel: C₆H₈BNO₂·HCl
MW: 173,40 g/mol
CAS nummer: 2096333-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8e5096a5b038c57ed9469d27c5ab43eabd9b0cb&vid=e8e5096a5b038c57ed9469d27c5ab43eabd9b0cb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) 95.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR