Metoclopramide ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide

M2218-5G M2218-25G
TCIAM2218-5GEA 575 SEK
TCIAM2218-5G TCIAM2218-25G
Metoclopramide ≥98.0% (by GC, titration analysis)
Metoclopramide
Formel: C₁₄H₂₂ClN₃O₂
MW: 299,80 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00211338
CAS nummer: 364-62-5
EINECS: 206-662-9
Merck Index: 14,06143

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR