Matrine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, Matridin-15-one

M2120-1G M2120-200MG
TCIAM2120-1GEA 3675 SEK
TCIAM2120-1G TCIAM2120-200MG
Matrine ≥98.0% (by GC, titration analysis)
Matrine
Formel: C₁₅H₂₄N₂O
MW: 248,37 g/mol
Kokpunkt: 223°C
Storage Temperature: Kylskåp
MDL Number: MFCD00210527
CAS nummer: 519-02-8
Merck Index: 14,05759

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 83.0 to 87.0 deg-C
Specific rotation [a]20/D +31.0 to +34.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR