N-Methyl-3-pyrrolidinol ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 3-Hydroxy-1-methylpyrrolidine, 1-Methyl-3-pyrrolidinol

M2115-5G M2115-25G
TCIAM2115-5GEA 990 SEK
TCIAM2115-5G TCIAM2115-25G
N-Methyl-3-pyrrolidinol ≥97.0%
N-Methyl-3-pyrrolidinol
Formel: C₅H₁₁NO
MW: 101,15 g/mol
Kokpunkt: 75°C
Flampunkt: 71°C
MDL Number: MFCD00003176
CAS nummer: 13220-33-2
EINECS: 236-189-3

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR