ɑ-Methyl-L-proline ≥96.0%
Leverantör: TCI EUROPE
Synonyms:
(S)-2-Methylpyrrolidine-2-carboxylic acid, H-α-Me-Pro-OH
M2077-1G
M2077-5G
TCIAM2077-1GEA
1875
SEK
TCIAM2077-1G
TCIAM2077-5G
ɑ-Methyl-L-proline ≥96.0%
alpha-Methyl-L-proline
Formel:
C₆H₁₁NO₂ MW: 129,16 g/mol Smältpunkt: 310°C |
MDL Number:
MFCD01318647 CAS nummer: 42856-71-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Specific rotation [a]20/D | -69.0 to -77.0 deg (C=1, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...