Methyl-2-(bromomethyl)acrylate ≥97.0%
Leverantör: TCI
Varning
M2067-1G M2067-5G
TCIAM2067-1GEA 350 SEK
TCIAM2067-1G TCIAM2067-5G
Methyl-2-(bromomethyl)acrylate ≥97.0%
Methyl-2-(bromomethyl)acrylate
Formel: C₅H₇BrO₂
Kokpunkt: 42°C
Flampunkt: 78°C
Storage Temperature: Kylskåp
MDL Number: MFCD00011697
CAS nummer: 4224-69-5
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %