4-Methoxybenzyl 2,2,2-trichloroacetimidate ≥96.0%
Leverantör: TCI
Varning
Synonyms: 2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
M2016-5G M2016-25G
TCIAM2016-5GEA 1200 SEK
TCIAM2016-5G TCIAM2016-25G
4-Methoxybenzyl 2,2,2-trichloroacetimidate ≥96.0%
4-Methoxybenzyl 2,2,2-trichloroacetimidate
Formel: C₁₀H₁₀Cl₃NO₂
Kokpunkt: 137°C
Storage Temperature: Frys
MDL Number: MFCD00134547
CAS nummer: 89238-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=642ff23f6868a36c9b115c0cd3cfd31694982f65&vid=642ff23f6868a36c9b115c0cd3cfd31694982f65&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %