2-(2-Methoxyphenoxy)ethylamine ≥95.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 1-(2-Aminoethoxy)-2-methoxybenzene
M2004-25G M2004-5G
TCIAM2004-25GEA 1675 SEK
TCIAM2004-25G TCIAM2004-5G
2-(2-Methoxyphenoxy)ethylamine ≥95.0% (by GC, titration analysis)
2-(2-Methoxyphenoxy)ethylamine
Formel: C₉H₁₃NO₂
MW: 167,21 g/mol
Kokpunkt: 98°C
MDL Number: MFCD00235185
CAS nummer: 1836-62-0
Order Now
Specifikation testresultat
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %