Methyl cis-4-aminocyclohexanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Leverantör: TCI EUROPE

Synonyms: cis-Methyl 4-aminocyclohexanecarboxylate hydrochloride, cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

M1985-5G M1985-1G
TCIAM1985-5GEA 2800 SEK
TCIAM1985-5G TCIAM1985-1G
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride
Formel: C₈H₁₅NO₂
MW: 193,67 g/mol
Smältpunkt: 188°C
MDL Number: MFCD08274537
CAS nummer: 61367-16-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR