1-Methyl-1,2-epoxycyclohexane ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Methyl-7-oxabicyclo[4.1.0]heptane, 1,2-Epoxy-1-methylcyclohexane
M1983-1G M1983-5G
TCIAM1983-1GEA 990 SEK
TCIAM1983-1G TCIAM1983-5G
1-Methyl-1,2-epoxycyclohexane ≥95.0%
1-Methyl-1,2-epoxycyclohexane
Formel: C₇H₁₂O
MW: 112,17 g/mol
Kokpunkt: 138°C
MDL Number: MFCD11846121
CAS nummer: 1713-33-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39ff5f69ec31ddfe3217cc103caf07fded0dbad2&vid=39ff5f69ec31ddfe3217cc103caf07fded0dbad2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %