1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine ≥96.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline, 4-(4-(4-Methoxyphenyl)piperazin-1-yl)aniline

M1920-25G M1920-5G
TCIAM1920-25GEA 2575 SEK
TCIAM1920-25G TCIAM1920-5G
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine ≥96.0% (by HPLC, titration analysis)
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
Formel: C₁₇H₂₁N₃O
MW: 283,38 g/mol
MDL Number: MFCD02071496
CAS nummer: 74852-62-3

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR