Methyl tridecafluoroheptanoate ≥96.0%

Leverantör: TCI
Varning

M1914-25G M1914-5G
TCIAM1914-25GEA 2225 SEK
TCIAM1914-25G TCIAM1914-5G
Methyl tridecafluoroheptanoate ≥96.0%
Methyl tridecafluoroheptanoate
Formel: C₈H₃F₁₃O₂
Kokpunkt: 141°C
MDL Number: MFCD00153212
CAS nummer: 14312-89-1
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR