(2-Mercaptoethyl)pyrazine ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-(Pyrazin-2-yl)ethanethiol, Pyrazineethanethiol
M1845-1G M1845-5G
TCIAM1845-1GEA 455 SEK
TCIAM1845-1G TCIAM1845-5G
(2-Mercaptoethyl)pyrazine ≥98.0%
(2-Mercaptoethyl)pyrazine
Formel: C₆H₈N₂S
MW: 140,20 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00053160
CAS nummer: 35250-53-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4987924ffd37c59a88c7f7ac45b8ee83769bfd39&vid=4987924ffd37c59a88c7f7ac45b8ee83769bfd39&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %