4'-Methoxy-N-methylformanilide ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
N-(4-Methoxyphenyl)-N-methylformamide
M1742-1G
M1742-5G
TCIAM1742-1GEA
1350
SEK
TCIAM1742-1G
TCIAM1742-5G
4'-Methoxy-N-methylformanilide ≥97.0%
4'-Methoxy-N-methylformanilide
Formel:
C₉H₁₁NO₂ MW: 165,19 g/mol |
MDL Number:
MFCD09038534 CAS nummer: 5279-51-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 97.0 % |
Freezing point | 22.0 to 27.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...