4-Methoxyphenyl-3-O-benzyl-β-D-galactopyranoside ≥98.0%

Leverantör: TCI EUROPE

M1725-1G M1725-5G
TCIAM1725-1GEA 1625 SEK
TCIAM1725-1G TCIAM1725-5G
4-Methoxyphenyl-3-O-benzyl-β-D-galactopyranoside ≥98.0%
4-Methoxyphenyl-3-O-benzyl-beta-D-galactopyranoside
Formel: C₂₀H₂₄O₇
MW: 376,41 g/mol
Storage Temperature: Kylskåp
CAS nummer: 383905-60-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR