Methanedisulphonyl dichloride ≥96.0%
Leverantör: TCI
|
Fara
|
M1669-25G
M1669-5G
TCIAM1669-25GEA
6900
SEK
TCIAM1669-25G
TCIAM1669-5G
Methanedisulphonyl dichloride ≥96.0%
Methanedisulphonyl dichloride
Formel:
CH₂Cl₂O₄S₂ Kokpunkt: 138°C |
MDL Number:
MFCD00176854 CAS nummer: 5799-68-8 EINECS: 227-350-9 UN: 3265 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (Argentmetric Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...