7-Methyl-1,4-benzothiazin-3-one
Leverantör: TCI EUROPE
M1661-5G
M1661-1G
TCIAM1661-5GEA
1450
SEK
TCIAM1661-5G
TCIAM1661-1G
7-Methyl-1,4-benzothiazin-3-one
7-Methyl-1,4-benzothiazin-3-one
Formel:
C₉H₉NOS MW: 179,24 g/mol Smältpunkt: 206°C |
MDL Number:
MFCD02660690 CAS nummer: 30322-02-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...