4-Methoxyphenyl-3-O-allyl-6-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0%
Leverantör: TCI
M1604-1G
M1604-5G
TCIAM1604-1GEA
925
SEK
TCIAM1604-1G
TCIAM1604-5G
4-Methoxyphenyl-3-O-allyl-6-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0%
4-Methoxyphenyl-3-O-allyl-6-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside
Formel:
C₃₁H₃₁NO₈ MW: 545.59 g/mol |
CAS nummer:
1820583-64-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...