4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98.0% (by GC, titration analysis) free radical

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, PTMA Monomer, 4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical, 4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate

M1531-1G M1531-5G
TCIAM1531-1GEA 1050 SEK
TCIAM1531-1G TCIAM1531-5G
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98.0% (by GC, titration analysis) free radical
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl
Formel: C₁₃H₂₂NO₃
MW: 240,32 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD17677357
CAS nummer: 15051-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbcf08bbbb19a9b02c92fa8604fecd991c2966ad&vid=fbcf08bbbb19a9b02c92fa8604fecd991c2966ad&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR