Methyltributylphosphonium iodide ≥98.0%

Leverantör: TCI
Varning

Synonyms: Methyltributylphosphoniumiodide

M1455-25G M1455-5G
TCIAM1455-25GEA 1150 SEK
TCIAM1455-25G TCIAM1455-5G
Methyltributylphosphonium iodide ≥98.0%
Methyltributylphosphonium iodide
Formel: C₁₃H₃₀IP
MDL Number: MFCD00031626
CAS nummer: 1702-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64dd94314795ceda7750f10f631db5c764a72811&vid=64dd94314795ceda7750f10f631db5c764a72811&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR