5-Methyluridine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

M1405-1G M1405-25G M1405-5G
TCIAM1405-1GEA 215 SEK
TCIAM1405-1G TCIAM1405-25G TCIAM1405-5G
5-Methyluridine ≥98.0% (by HPLC, titration analysis)
5-Methyluridine
Formel: C₁₀H₁₄N₂O₆
MW: 258,23 g/mol
MDL Number: MFCD00006535
CAS nummer: 1463-10-1
EINECS: 215-973-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Melting point 182.0 to 186.0 deg-C
Specific rotation [a]20/D -7.0 to -10.0 deg (C=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR