5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin ≥90.0%
Leverantör: TCI EUROPE
Synonyms:
5-(4-Carbomethoxyphenyl)-10,15,20-triphenylporphyrin, 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenyl-21H,23H-porphine
M1338-1G
M1338-100MG
TCIAM1338-1GEA
7925
SEK
TCIAM1338-1G
TCIAM1338-100MG
5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin ≥90.0%
5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin
Formel:
C₄₆H₃₂N₄O₂ MW: 672,79 g/mol |
MDL Number:
MFCD02093480 CAS nummer: 119730-06-2 |
Specifikation testresultat
Appearance | Dark red to Dark purple to Dark blue powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Lambda max. | 411.0 to 415.0 nm (methanol) |
Absorbance (E1%1cm) | min. 6000 (methanol, 411 to 415nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...