Furfuryl methacrylate ≥95.0% stabilised

Leverantör: TCI
Varning

Synonyms: Methacrylic Acid Furfuryl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M1335-25G M1335-5G
TCIAM1335-25GEA 2175 SEK
TCIAM1335-25G TCIAM1335-5G
Furfuryl methacrylate ≥95.0% stabilised
Furfuryl methacrylate
Formel: C₉H₁₀O₃
Kokpunkt: 82°C
Flampunkt: 90°C
MDL Number: MFCD00046113
CAS nummer: 3454-28-2
EINECS: 222-383-5

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR