Methyl-2-methoxyphenylacetate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-Methoxyphenylacetic Acid Methyl Ester, Methyl 2-Methoxyphenylacetate

M1329-25G M1329-5G
TCIAM1329-25GEA 1625 SEK
TCIAM1329-25G TCIAM1329-5G
Methyl-2-methoxyphenylacetate ≥98.0%
Methyl-2-methoxyphenylacetate
Formel: C₁₀H₁₂O₃
MW: 180,20 g/mol
MDL Number: MFCD02093479
CAS nummer: 27798-60-3
EINECS: 248-662-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d039856902a3278fce801da624e54b81bcb7baf&vid=4d039856902a3278fce801da624e54b81bcb7baf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR