Cyclohexylacetic acid 98+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Cyclohexaneacetic acid
111270050 111270250 111271000
ACRO111270050EA 645 SEK
ACRO111270050 ACRO111270250 ACRO111271000
Cyclohexylacetic acid 98+%
Cyclohexylacetic acid
Formel: C₈H₁₄O₂
Kokpunkt: 242…244 °C (760 mmHg)
Smältpunkt: 28…32 °C
Densitet: 1,007 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001518
CAS nummer: 5292-21-7
EINECS: 226-132-0
Order Now
Specifikation testresultat
Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 27 to 31 °C
Titration with NaOH ≥98.0 %
Identification Passes test
Sensitivity (to amino acids) Passes test