2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine

M1245-25G M1245-5G
TCIAM1245-25GEA 1650 SEK
TCIAM1245-25G TCIAM1245-5G
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0%
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Formel: C₁₂H₇Cl₆N₃O
MW: 421,91 g/mol
MDL Number: MFCD00521486
CAS nummer: 3584-23-4
EINECS: 222-711-7

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 143.0 to 147.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR