(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: (+/-)-Naproxen, 2-(6-Methoxynaphthalen-2-yl)propanoic acid, (+/-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid

M1220-25G M1220-5G
TCIAM1220-25GEA 2500 SEK
TCIAM1220-25G TCIAM1220-5G
(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98.0% (by GC, titration analysis)
(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid
Formel: C₁₄H₁₄O₃
MW: 230,26 g/mol
MDL Number: MFCD00439456
CAS nummer: 23981-80-8
EINECS: 245-969-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6efc0b3d97c7d4eb2540c5be4b0bd3a6f0076e57&vid=6efc0b3d97c7d4eb2540c5be4b0bd3a6f0076e57&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 154.0 to 158.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR