4-Methyl-1-acetonaphthone ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Acetyl-4-methylnaphthalene, 1-(4-Methylnaphthalen-1-yl)ethanone

M1202-25G M1202-5G
TCIAM1202-25GEA 2550 SEK
TCIAM1202-25G TCIAM1202-5G
4-Methyl-1-acetonaphthone ≥97.0%
4-Methyl-1-acetonaphthone
Formel: C₁₃H₁₂O
MW: 184,24 g/mol
Kokpunkt: 136°C
MDL Number: MFCD00671556
CAS nummer: 28418-86-2

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 97.0 %
Melting point 35.0 to 39.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR