Methyl-3-methoxyisobutyrate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Methoxyisobutyric Acid Methyl Ester, Methyl 3-Methoxy-2-methylpropionate, 3-Methoxy-2-methylpropionic Acid Methyl Ester
M1191-25ML
M1191-500ML
TCIAM1191-25MLEA
230
SEK
TCIAM1191-25ML
TCIAM1191-500ML
Methyl-3-methoxyisobutyrate ≥98.0%
Methyl-3-methoxyisobutyrate
Formel:
C₆H₁₂O₃ MW: 132,16 g/mol Kokpunkt: 148°C Flampunkt: 46°C |
MDL Number:
MFCD00671553 CAS nummer: 3852-11-7 EINECS: 223-359-7 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...