2,4-Mesitylenedisulphonyl dichloride ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2,4,6-Trimethyl-1,3-benzenedisulphonyl Dichloride, 2,4,6-Trimethylbenzene-1,3-disulphonyl dichloride
M1186-25G
M1186-5G
TCIAM1186-25GEA
7625
SEK
TCIAM1186-25G
TCIAM1186-5G
2,4-Mesitylenedisulphonyl dichloride ≥98.0%
2,4-Mesitylenedisulphonyl dichloride
Formel:
C₉H₁₀Cl₂O₄S₂ MW: 317,20 g/mol |
MDL Number:
MFCD00009796 CAS nummer: 68985-08-0 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...