3-Methylpyrrole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Methyl-1H-pyrrole

M1144-1G M1144-5G
TCIAM1144-1GEA 2425 SEK
TCIAM1144-1G TCIAM1144-5G
3-Methylpyrrole ≥98.0%
3-Methylpyrrole
Formel: C₅H₇N
MW: 81,12 g/mol
Kokpunkt: 144°C
MDL Number: MFCD00083419
CAS nummer: 616-43-3
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR