(3R)-(+)-3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: (3R)-(+)-3-(Methylamino)pyrrolidine dihydrochloride
M1107-1G M1107-5G
TCIAM1107-1GEA 2125 SEK
TCIAM1107-1G TCIAM1107-5G
(3R)-(+)-3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
(3R)-(+)-3-(Methylamino)pyrrolidine
Formel: C₅H₁₂N₂
MW: 100,17 g/mol
Kokpunkt: 164°C
Storage Temperature: Kylskåp
MDL Number: MFCD00191461
CAS nummer: 139015-33-1
UN: 2734
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +17.0 to +22.0 deg (C=10, EtOH)