N-Methyl-N,O-bis(trimethylsilyl)hydroxylamine

Leverantör: TCI EUROPE
Fara

M1077-1ML M1077-5ML
TCIAM1077-1MLEA 1475 SEK
TCIAM1077-1ML TCIAM1077-5ML
N-Methyl-N,O-bis(trimethylsilyl)hydroxylamine
N-Methyl-N,O-bis(trimethylsilyl)hydroxylamine
Formel: C₇H₂₁NOSi₂
Storage Temperature: Kylskåp
MDL Number: MFCD00191463
CAS nummer: 22737-33-3
UN: 2920
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR