1-(2-Methoxyphenoxy)-2-propanol ≥90.0%

Leverantör: TCI

M1060-25G M1060-5G
TCIAM1060-25GEA 1750 SEK
TCIAM1060-25G TCIAM1060-5G
1-(2-Methoxyphenoxy)-2-propanol ≥90.0%
1-(2-Methoxyphenoxy)-2-propanol
Formel: C₁₀H₁₄O₃
MDL Number: MFCD00191539
CAS nummer: 64120-49-6

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR